The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168325.1 | NP_001161797.1 | 5 - 6 | 601 | 99 |
NM_017899.3 | NP_060369.3 | 6 - 7 | 682 | 99 | |
NR_031766.2 | 6 - 7 | 687 | 99 | ||
XM_011538503.1 | XP_011536805.1 | 6 - 7 | 497 | 99 | |
GenBank mRNA | AF443207.1 | - | 6 - 7 | 639 | 99 |
AI910454.1 | - | 2 - 3 | 99 | ||
AK000614.1 | - | 6 - 7 | 730 | 99 | |
BC015221.1 | - | 6 - 7 | 639 | 99 | |
BE670015.1 | - | 2 - 3 | 99 | ||
BM666793.1 | - | 2 - 3 | 99 | ||
BQ434057.1 | - | 6 - 7 | 620 | 99 | |
BQ949772.1 | - | 5 - 6 | 555 | 99 |