Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EGLN2
- Entrez Gene ID
-
112398
- Gene Name
- egl-9 family hypoxia inducible factor 2
- Gene Aliases
-
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
- Chromosome Location
-
Chr.19: 40799143 - 40808441 on Build GRCh38
- Gene Symbol
- RAB4B-EGLN2
- Entrez Gene ID
-
100529264
- Gene Name
- RAB4B-EGLN2 readthrough (NMD candidate)
- Gene Aliases
-
RERT-lncRNA
- Chromosome Location
-
Chr.19: 40778219 - 40808441 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037791.1 |
|
8 - 9 |
1890 |
64 |
GenBank mRNA |
AK291385.1 |
- |
8 - 9 |
1838 |
64 |
View Full Product Details