Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP2
Entrez Gene ID
7077
Gene Name
TIMP metallopeptidase inhibitor 2
Gene Aliases
CSC-21K, DDC8
Chromosome Location
Chr.17: 78852977 - 78925390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003255.4 NP_003246.1 2 - 3 537 146
GenBank mRNA AK294290.1 - 2 - 3 297 146
AL713764.1 - 1 - 2 322 146
BC039613.1 - 1 - 2 275 146
BC040039.1 - 1 - 2 187 146
BC040445.1 - 1 - 2 272 146
BC052605.1 - 2 - 3 514 146
BC071586.1 - 2 - 3 534 146
J05593.1 - 2 - 3 505 146
M32304.1 - 2 - 3 489 146
S48568.1 - 2 - 3 505 146
X54533.1 - 2 - 3 148 146

View Full Product Details