The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300916.1 | NP_001287845.1 | 1 - 2 | 293 | 130 |
NM_182948.3 | NP_891993.1 | 1 - 2 | 293 | 130 | |
XM_005271020.1 | XP_005271077.1 | 1 - 2 | 273 | 130 | |
GenBank mRNA | AK304322.1 | - | 1 - 2 | 218 | 130 |
BM905428.1 | - | 1 - 2 | 175 | 130 | |
BX485407.1 | - | 1 - 2 | 274 | 130 | |
BX537705.1 | - | 1 - 2 | 270 | 130 | |
BX641026.1 | - | 1 - 2 | 269 | 130 |