The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321001.1 | NP_001307930.1 | 3 - 4 | 264 | 66 |
NM_001321002.1 | NP_001307931.1 | 3 - 4 | 236 | 66 | |
NM_001321004.1 | NP_001307933.1 | 2 - 3 | 498 | 66 | |
NM_001321005.1 | NP_001307934.1 | 3 - 4 | 308 | 66 | |
NM_002453.2 | NP_002444.2 | 3 - 4 | 308 | 66 | |
XM_011532871.2 | XP_011531173.1 | 2 - 3 | 141 | 66 | |
XM_017004164.1 | XP_016859653.1 | 3 - 4 | 302 | 66 | |
GenBank mRNA | AF494407.1 | - | 3 - 4 | 288 | 66 |
AK225184.1 | - | 3 - 4 | 277 | 66 | |
AK307993.1 | - | 3 - 4 | 288 | 66 | |
BC017243.1 | - | 3 - 4 | 256 | 66 | |
BC039848.1 | - | 3 - 4 | 266 | 66 | |
BC054048.1 | - | 3 - 4 | 205 | 66 | |
BC064977.1 | - | 3 - 4 | 248 | 66 | |
BP282053.1 | - | 3 - 4 | 216 | 66 | |
CB961193.1 | - | 3 - 4 | 335 | 66 | |
DB478758.2 | - | 2 - 3 | 467 | 66 | |
L34600.1 | - | 3 - 4 | 262 | 66 |