Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
Chromosome Location
Chr.X: 131273506 - 131289457 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170961.1 NP_001164432.1 18 - 19 4052 118
NM_001170962.1 NP_001164433.1 17 - 18 4010 118
NM_001555.4 NP_001546.2 18 - 19 4037 118
XM_011531330.1 XP_011529632.1 19 - 20 3950 118
XM_011531333.1 XP_011529635.1 18 - 19 3803 118
XM_011531334.1 XP_011529636.1 16 - 17 3476 118
GenBank mRNA AF034198.1 - 17 - 18 3859 118
AK226008.1 - 18 - 19 3909 118
AK295438.1 - 9 - 10 2088 118
AK307871.1 - 18 - 19 3913 118
AL137369.1 - 2 - 3 299 118
BC063884.1 - 18 - 19 3873 118
Y10523.1 - 17 - 18 3834 118

View Full Product Details