The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003968.3 | NP_003959.3 | 16 - 17 | 1268 | 79 |
NM_198195.1 | NP_937838.1 | 15 - 16 | 1226 | 79 | |
XM_011534210.1 | XP_011532512.1 | 15 - 16 | 1182 | 79 | |
XM_011534211.1 | XP_011532513.1 | 14 - 15 | 1135 | 79 | |
GenBank mRNA | AB012190.1 | - | 14 - 15 | 1185 | 79 |
AF046024.1 | - | 15 - 16 | 1269 | 79 | |
AK002159.1 | - | 15 - 16 | 1213 | 79 | |
AK289392.1 | - | 16 - 17 | 1254 | 79 | |
AK298878.1 | - | 11 - 12 | 941 | 79 | |
AK314751.1 | - | 16 - 17 | 1251 | 79 | |
AK316540.1 | - | 13 - 14 | 1105 | 79 | |
AL117566.1 | - | 16 - 17 | 1264 | 79 | |
BC022853.1 | - | 16 - 17 | 1267 | 79 | |
CR533537.1 | - | 16 - 17 | 1248 | 79 |