The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135947.2 | NP_001129419.1 | 1 - 2 | 96 | 88 |
NM_001265582.1 | NP_001252511.1 | 1 - 2 | 96 | 88 | |
NM_030914.3 | NP_112176.1 | 1 - 2 | 96 | 88 | |
GenBank mRNA | AK001880.1 | - | 1 - 2 | 45 | 88 |
AK097029.1 | - | 1 - 2 | 42 | 88 | |
AK300877.1 | - | 1 - 2 | 43 | 88 | |
BC003581.1 | - | 1 - 2 | 54 | 88 |