Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINS1
Entrez Gene ID
55180
Gene Name
lines homolog 1
Gene Aliases
LINS, MRT27, WINS1
Chromosome Location
Chr.15: 100566914 - 100602440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040616.2 NP_001035706.1 1 - 2 119 122
GenBank mRNA AK001445.1 - 1 - 2 53 122
AK292972.1 - 1 - 2 50 122
AK298946.1 - 1 - 2 55 122
DA447859.1 - 1 - 2 119 122
DA674923.1 - 1 - 2 78 122
DA880742.1 - 1 - 2 63 122

View Full Product Details