The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004526.3 | NP_004517.2 | 5 - 6 | 1006 | 88 |
NR_073375.1 | 5 - 6 | 1025 | 88 | ||
GenBank mRNA | AK128291.1 | - | 5 - 6 | 665 | 88 |
AK299934.1 | - | 5 - 6 | 972 | 88 | |
AK304204.1 | - | 6 - 7 | 877 | 88 | |
AK309335.1 | - | 6 - 7 | 917 | 88 | |
BC006165.1 | - | 5 - 6 | 922 | 88 | |
BC007670.2 | - | 5 - 6 | 944 | 88 | |
BC007938.2 | - | 5 - 6 | 914 | 88 | |
BC014272.2 | - | 5 - 6 | 945 | 88 | |
BC017258.2 | - | 5 - 6 | 934 | 88 | |
BC017490.1 | - | 5 - 6 | 911 | 88 | |
BC030131.2 | - | 3 - 4 | 608 | 88 | |
BT009734.1 | - | 4 - 5 | 869 | 88 | |
D21063.1 | - | 5 - 6 | 927 | 88 | |
D83987.1 | - | 4 - 5 | 893 | 88 | |
X67334.1 | - | 4 - 5 | 893 | 88 |