The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083585.2 | NP_001077054.1 | 11 - 12 | 2006 | 76 |
NM_001291581.1 | NP_001278510.1 | 10 - 11 | 1877 | 76 | |
NM_004703.5 | NP_004694.2 | 11 - 12 | 2006 | 76 | |
XM_011524060.1 | XP_011522362.1 | 12 - 13 | 2072 | 76 | |
XM_017025294.1 | XP_016880783.1 | 10 - 11 | 1599 | 76 | |
XM_017025295.1 | XP_016880784.1 | 9 - 10 | 1533 | 76 | |
GenBank mRNA | AF098638.1 | - | 11 - 12 | 1782 | 76 |
AK297535.1 | - | 10 - 11 | 1866 | 76 | |
AK310932.1 | - | 10 - 11 | 1818 | 76 | |
AK314183.1 | - | 11 - 12 | 1997 | 76 | |
BC032435.1 | - | 11 - 12 | 1960 | 76 | |
BC041700.1 | - | 11 - 12 | 1972 | 76 | |
X77723.1 | - | 11 - 12 | 1855 | 76 | |
X91141.1 | - | 11 - 12 | 1970 | 76 |