Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN2L
Entrez Gene ID
81562
Gene Name
lectin, mannose binding 2 like
Gene Aliases
MRT52, VIPL
Chromosome Location
Chr.2: 96705929 - 96740092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142292.1 NP_001135764.1 1 - 2 227 70
NM_001322346.1 NP_001309275.1 1 - 2 243 70
NM_001322352.1 NP_001309281.1 1 - 2 243 70
NM_001322354.1 NP_001309283.1 1 - 2 84
NM_001322355.1 NP_001309284.1 1 - 2 243 70
NM_001322356.1 NP_001309285.1 1 - 2 243 70
NM_030805.3 NP_110432.1 1 - 2 227 70
GenBank mRNA AJ549957.1 - 1 - 2 191 70
AK075347.1 - 1 - 2 215 70
AK222828.1 - 1 - 2 205 70
AK299740.1 - 1 - 2 215 70
AK308085.1 - 1 - 2 214 70
AK310100.1 - 1 - 2 244 70
AK316331.1 - 1 - 2 215 70
AL136617.1 - 1 - 2 219 70
AY358929.1 - 1 - 2 198 70
BC000347.2 - 1 - 2 190 70
BC005822.2 - 1 - 2 190 70
BC005862.2 - 1 - 2 190 70
BC067265.1 - 1 - 2 197 70
BX648002.1 - 1 - 2 205 70
DA521101.1 - 1 - 2 204 70
DC354352.1 - 1 - 2 84

View Full Product Details