Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNL1
Entrez Gene ID
57018
Gene Name
cyclin L1
Gene Aliases
ANIA6A, BM-001, PRO1073, ania-6a
Chromosome Location
Chr.3: 157146502 - 157160760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005247648.3 XP_005247705.1 3 - 4 541 58
XM_017006891.1 XP_016862380.1 3 - 4 572 58
XM_017006892.1 XP_016862381.1 3 - 4 589 58
XM_017006893.1 XP_016862382.1 3 - 4 572 58
XM_017006894.1 XP_016862383.1 3 - 4 589 58
XM_017006895.1 XP_016862384.1 3 - 4 541 58
XM_017006897.1 XP_016862386.1 3 - 4 541 58
GenBank mRNA AF367477.1 - 3 - 4 491 58
AY034790.1 - 3 - 4 831 58
BC038394.1 - 3 - 4 532 58

View Full Product Details