Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNL1
Entrez Gene ID
57018
Gene Name
cyclin L1
Gene Aliases
ANIA6A, BM-001, PRO1073, ania-6a
Chromosome Location
Chr.3: 157146502 - 157160760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005247648.3 XP_005247705.1 4 - 5 613 113
XM_011513011.2 XP_011511313.1 4 - 5 1165 113
XM_017006893.1 XP_016862382.1 4 - 5 644 113
XM_017006895.1 XP_016862384.1 4 - 5 613 113
GenBank mRNA AY034790.1 - 4 - 5 903 113
BC038394.1 - 4 - 5 604 113

View Full Product Details