Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELN
Entrez Gene ID
2006
Gene Name
elastin
Gene Aliases
SVAS, WBS, WS
Chromosome Location
Chr.7: 74027772 - 74069907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000501.3 NP_000492.2 8 - 9 828 128
NM_001081752.2 NP_001075221.1 7 - 8 798 128
NM_001081753.2 NP_001075222.1 8 - 9 843 128
NM_001081754.2 NP_001075223.1 8 - 9 843 128
NM_001081755.2 NP_001075224.1 8 - 9 828 128
NM_001278912.1 NP_001265841.1 8 - 9 828 128
NM_001278913.1 NP_001265842.1 7 - 8 792 128
NM_001278914.1 NP_001265843.1 7 - 8 813 128
NM_001278915.1 NP_001265844.1 8 - 9 828 128
NM_001278916.1 NP_001265845.1 8 - 9 828 128
NM_001278917.1 NP_001265846.1 7 - 8 798 128
NM_001278939.1 NP_001265868.1 8 - 9 828 128
XM_005250187.1 XP_005250244.1 7 - 8 469 128
XM_005250188.1 XP_005250245.1 8 - 9 503 128
XM_011515868.1 XP_011514170.1 8 - 9 529 128
XM_011515869.1 XP_011514171.1 7 - 8 458 128
XM_011515870.1 XP_011514172.1 7 - 8 824 128
XM_011515871.1 XP_011514173.1 8 - 9 529 128
XM_011515872.1 XP_011514174.1 8 - 9 529 128
XM_011515873.1 XP_011514175.1 8 - 9 529 128
XM_011515874.1 XP_011514176.1 6 - 7 794 128
XM_011515875.1 XP_011514177.1 6 - 7 450 128
XM_011515876.1 XP_011514178.1 8 - 9 529 128
XM_011515877.1 XP_011514179.1 8 - 9 529 128
XM_017011813.1 XP_016867302.1 7 - 8 414 128
XM_017011814.1 XP_016867303.1 8 - 9 529 128
GenBank mRNA AB208942.1 - 7 - 8 424 128
AK075554.1 - 7 - 8 475 128
AK094138.1 - 7 - 8 351 128
AK095990.1 - 8 - 9 463 128
AK125511.1 - 7 - 8 473 128
AK225659.1 - 8 - 9 442 128
AK304845.1 - 8 - 9 828 128
BC035570.1 - 8 - 9 433 128
BC050379.1 - 8 - 9 446 128
BC065566.1 - 8 - 9 459 128
BX537939.1 - 8 - 9 463 128
BX538199.1 - 7 - 8 430 128
M36860.1 - 8 - 9 478 128
X52896.1 - 8 - 9 477 128

View Full Product Details