The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145023.1 | NP_001138495.1 | 5 - 6 | 901 | 90 |
NM_138355.3 | NP_612364.2 | 5 - 6 | 901 | 90 | |
XM_005257776.4 | XP_005257833.1 | 5 - 6 | 881 | 90 | |
XM_017025286.1 | XP_016880775.1 | 5 - 6 | 829 | 90 | |
GenBank mRNA | AK290646.1 | - | 5 - 6 | 804 | 90 |
AK303835.1 | - | 5 - 6 | 857 | 90 | |
BC002980.1 | - | 5 - 6 | 844 | 90 | |
BC010408.2 | - | 5 - 6 | 815 | 90 | |
BC017317.2 | - | 5 - 6 | 814 | 90 |