The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281463.1 | NP_001268392.1 | 18 - 19 | 2983 | 106 |
NM_006306.3 | NP_006297.2 | 17 - 18 | 2837 | 106 | |
GenBank mRNA | BC112127.1 | - | 17 - 18 | 2777 | 106 |
BC171726.1 | - | 18 - 19 | 2923 | 106 | |
D80000.2 | - | 17 - 18 | 2709 | 106 | |
S78271.1 | - | 17 - 18 | 2742 | 106 |