The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281463.1 | NP_001268392.1 | 12 - 13 | 2183 | 87 |
NM_006306.3 | NP_006297.2 | 11 - 12 | 2037 | 87 | |
GenBank mRNA | AK291921.1 | - | 11 - 12 | 1996 | 87 |
AK301266.1 | - | 8 - 9 | 1261 | 87 | |
BC064368.1 | - | 11 - 12 | 1968 | 87 | |
BC112127.1 | - | 11 - 12 | 1977 | 87 | |
BC171726.1 | - | 12 - 13 | 2123 | 87 | |
BX640922.1 | - | 11 - 12 | 2080 | 87 | |
D80000.2 | - | 11 - 12 | 1909 | 87 | |
S78271.1 | - | 11 - 12 | 1942 | 87 |