The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016930.2 | NP_058626.1 | 6 - 7 | 695 | 147 |
XM_011513480.1 | XP_011511782.1 | 7 - 8 | 767 | 147 | |
XM_011513483.1 | XP_011511785.1 | 7 - 8 | 727 | 147 | |
XM_011513485.2 | XP_011511787.1 | 6 - 7 | 622 | 147 | |
XM_017008297.1 | XP_016863786.1 | 7 - 8 | 1348 | 147 | |
GenBank mRNA | AB028741.1 | - | 6 - 7 | 614 | 147 |
AK310026.1 | - | 6 - 7 | 657 | 147 | |
AY453396.1 | - | 6 - 7 | 611 | 147 | |
BT020135.1 | - | 6 - 7 | 611 | 147 |