The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001884.3 | NP_001875.1 | 3 - 4 | 788 | 60 |
XM_011543168.2 | XP_011541470.1 | 3 - 4 | 722 | 60 | |
XM_017009051.1 | XP_016864540.1 | 4 - 5 | 971 | 60 | |
XM_017009052.1 | XP_016864541.1 | 3 - 4 | 584 | 60 | |
XM_017009053.1 | XP_016864542.1 | 3 - 4 | 768 | 60 | |
XM_017009054.1 | XP_016864543.1 | 3 - 4 | 723 | 60 | |
GenBank mRNA | AK313713.1 | - | 3 - 4 | 743 | 60 |
BC057808.1 | - | 2 - 3 | 544 | 60 | |
U43328.1 | - | 3 - 4 | 590 | 60 | |
X17405.1 | - | 3 - 4 | 788 | 60 | |
X78076.1 | - | 3 - 4 | 788 | 60 |