Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMPD2
Entrez Gene ID
271
Gene Name
adenosine monophosphate deaminase 2
Gene Aliases
PCH9, SPG63
Chromosome Location
Chr.1: 109619813 - 109632055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257360.1 NP_001244289.1 9 - 10 1675 56
NM_001257361.1 NP_001244290.1 8 - 9 958 56
NM_001308170.1 NP_001295099.1 7 - 8 1078 56
NM_004037.7 NP_004028.3 8 - 9 1542 56
NM_139156.3 NP_631895.1 8 - 9 1322 56
NM_203404.1 NP_981949.1 6 - 7 960 56
GenBank mRNA AK025706.1 - 8 - 9 1298 56
AK296394.1 - 8 - 9 914 56
AK302939.1 - 8 - 9 920 56
AK308907.1 - 9 - 10 1148 56
AK314494.1 - 9 - 10 1598 56
BC007711.2 - 8 - 9 1247 56
BC075844.1 - 8 - 9 1247 56
S47833.1 - 7 - 8 933 56
U16270.1 - 3 - 4 346 56

View Full Product Details