The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319210.1 | NP_001306139.1 | 10 - 11 | 1466 | 81 |
NM_001319211.1 | NP_001306140.1 | 10 - 11 | 1098 | 81 | |
NM_001319212.1 | NP_001306141.1 | 9 - 10 | 1378 | 81 | |
NM_003672.3 | NP_003663.2 | 10 - 11 | 1466 | 81 | |
NM_033312.2 | NP_201569.1 | 10 - 11 | 1466 | 81 | |
NM_033313.2 | NP_201570.1 | 10 - 11 | 1466 | 81 | |
XM_005271294.3 | XP_005271351.1 | 10 - 11 | 1006 | 81 | |
XM_005271296.3 | XP_005271353.1 | 10 - 11 | 1003 | 81 | |
XM_011542340.2 | XP_011540642.1 | 10 - 11 | 1005 | 81 | |
XM_011542341.2 | XP_011540643.1 | 9 - 10 | 946 | 81 | |
XM_011542345.2 | XP_011540647.1 | 7 - 8 | 857 | 81 | |
XM_011542346.2 | XP_011540648.1 | 10 - 11 | 1014 | 81 | |
XM_017002646.1 | XP_016858135.1 | 10 - 11 | 1006 | 81 | |
XM_017002647.1 | XP_016858136.1 | 10 - 11 | 1547 | 81 | |
GenBank mRNA | AB209857.1 | - | 7 - 8 | 895 | 81 |
AF000367.1 | - | 10 - 11 | 1333 | 81 | |
AF064102.1 | - | 10 - 11 | 1441 | 81 | |
AF064103.1 | - | 10 - 11 | 1431 | 81 | |
AF122013.1 | - | 10 - 11 | 978 | 81 | |
AK302310.1 | - | 10 - 11 | 1087 | 81 | |
BC038979.1 | - | 9 - 10 | 81 | ||
BC093916.1 | - | 10 - 11 | 1073 | 81 | |
BC093918.1 | - | 10 - 11 | 1073 | 81 | |
DQ530256.1 | - | 10 - 11 | 984 | 81 |