The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321917.1 | NP_001308846.1 | 4 - 5 | 487 | 149 |
NM_133466.3 | NP_597723.1 | 4 - 5 | 484 | 149 | |
GenBank mRNA | AK074942.1 | - | 4 - 5 | 466 | 149 |
AK315709.1 | - | 4 - 5 | 444 | 149 | |
AL834267.1 | - | 4 - 5 | 458 | 149 | |
BC032577.2 | - | 4 - 5 | 451 | 149 | |
GU388429.1 | - | 4 - 5 | 454 | 149 |