The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001347.3 | NP_001338.2 | 19 - 20 | 2393 | 60 |
XM_011513411.1 | XP_011511713.1 | 19 - 20 | 2378 | 60 | |
GenBank mRNA | AB209503.1 | - | 19 - 20 | 2144 | 60 |
BC063801.1 | - | 19 - 20 | 2401 | 60 | |
GU727647.1 | - | 19 - 20 | 2331 | 60 | |
L38707.1 | - | 19 - 20 | 2353 | 60 |