Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCARF1
Entrez Gene ID
8578
Gene Name
scavenger receptor class F member 1
Gene Aliases
SREC, SREC-I, SREC1
Chromosome Location
Chr.17: 1633858 - 1645789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003693.3 NP_003684.2 4 - 5 887 163
NM_145350.2 NP_663325.1 4 - 5 887 163
NR_028075.2 4 - 5 887 163
NR_102409.1 4 - 5 924 163
GenBank mRNA AB052947.1 - 4 - 5 821 163
AB052948.1 - 4 - 5 854 163
AB052949.1 - 4 - 5 819 163
AB052950.1 - 4 - 5 811 163
AK291810.1 - 4 - 5 830 163
AK310174.1 - 4 - 5 847 163
AK311370.1 - 2 - 3 813 163
BC039735.1 - 4 - 5 855 163
D63483.1 - 4 - 5 797 163
D86864.1 - 4 - 5 821 163

View Full Product Details