The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020812.3 | NP_065863.2 | 29 - 30 | 3768 | 73 |
XM_005260000.2 | XP_005260057.1 | 31 - 32 | 3975 | 73 | |
XM_005260001.2 | XP_005260058.1 | 30 - 31 | 3882 | 73 | |
XM_006722804.3 | XP_006722867.1 | 8 - 9 | 1156 | 73 | |
XM_011528150.1 | XP_011526452.1 | 30 - 31 | 3915 | 73 | |
XM_011528151.1 | XP_011526453.1 | 30 - 31 | 3903 | 73 | |
XM_011528152.1 | XP_011526454.1 | 29 - 30 | 3810 | 73 | |
GenBank mRNA | AB037816.2 | - | 29 - 30 | 3728 | 73 |
AK295664.1 | - | 15 - 16 | 2032 | 73 | |
AK316063.1 | - | 13 - 14 | 1794 | 73 | |
BC008335.1 | - | 1 - 2 | 157 | 73 | |
BC146786.1 | - | 29 - 30 | 3728 | 73 | |
BI822534.1 | - | 3 - 4 | 479 | 73 | |
CX759906.1 | - | - | 175 | 73 |