The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020812.3 | NP_065863.2 | 39 - 40 | 5137 | 68 |
XM_005260000.2 | XP_005260057.1 | 41 - 42 | 5344 | 68 | |
XM_005260001.2 | XP_005260058.1 | 40 - 41 | 5251 | 68 | |
XM_006722804.3 | XP_006722867.1 | 18 - 19 | 2525 | 68 | |
XM_011528150.1 | XP_011526452.1 | 40 - 41 | 5284 | 68 | |
XM_011528151.1 | XP_011526453.1 | 40 - 41 | 5272 | 68 | |
XM_011528152.1 | XP_011526454.1 | 39 - 40 | 5179 | 68 | |
GenBank mRNA | AB037816.2 | - | 39 - 40 | 5097 | 68 |
AK295664.1 | - | 25 - 26 | 3401 | 68 | |
AK316063.1 | - | 23 - 24 | 3163 | 68 | |
BC008335.1 | - | 11 - 12 | 1526 | 68 | |
BC051330.1 | - | 4 - 5 | 472 | 68 | |
BC146786.1 | - | 39 - 40 | 5097 | 68 |