The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199141.1 | NP_954592.1 | 12 - 13 | 1553 | 108 |
XM_005259708.4 | XP_005259765.1 | 12 - 13 | 1519 | 108 | |
XM_011527638.2 | XP_011525940.1 | 11 - 12 | 1309 | 108 | |
GenBank mRNA | AF055027.1 | - | 7 - 8 | 728 | 108 |
AL833242.1 | - | 12 - 13 | 1346 | 108 | |
BC046240.1 | - | - | 850 | 108 |