Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCP2
Entrez Gene ID
3937
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
SLP-76, SLP76
Chromosome Location
Chr.5: 170248084 - 170297818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005565.3 NP_005556.1 5 - 6 493 103
GenBank mRNA AK292890.1 - 5 - 6 454 103
BC016618.1 - 5 - 6 430 103
BT007273.1 - 5 - 6 286 103
FN432830.1 - 5 - 6 286 183
U20158.1 - 5 - 6 541 103

View Full Product Details