The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145266.4 | NP_660309.1 | 2 - 3 | 325 | 114 |
GenBank mRNA | AK311957.1 | - | 2 - 3 | 288 | 114 |
BC017934.1 | - | 2 - 3 | 325 | 114 | |
BX538290.1 | - | 2 - 3 | 326 | 114 | |
FJ215684.1 | - | 2 - 3 | 265 | 114 |