Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
Chromosome Location
Chr.6: 24171755 - 24383292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195610.1 NP_001182539.1 5 - 6 766 112
NM_016356.4 NP_057440.2 4 - 5 859 112
GenBank mRNA AB032980.2 - 4 - 5 557 112
AF181721.1 - 4 - 5 859 112
AK027036.1 - 4 - 5 675 112
BC050704.1 - 5 - 6 766 112

View Full Product Details