The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317964.1 | NP_001304893.1 | 1 - 2 | 299 | 71 |
NM_016460.3 | NP_057544.2 | 1 - 2 | 299 | 71 | |
NM_020117.10 | NP_064502.9 | 1 - 2 | 299 | 71 | |
GenBank mRNA | AB037773.1 | - | 1 - 2 | 172 | 71 |
AF151026.1 | - | 1 - 2 | 139 | 71 | |
AK025441.1 | - | 1 - 2 | 135 | 71 | |
AK293750.1 | - | 1 - 2 | 129 | 71 | |
AK304134.1 | - | 1 - 2 | 115 | 71 | |
AK315178.1 | - | 1 - 2 | 72 | ||
BC068041.1 | - | 1 - 2 | 119 | 71 | |
BC105581.1 | - | 1 - 2 | 141 | 71 | |
BC150213.1 | - | 1 - 2 | 150 | 71 | |
BC151214.1 | - | 1 - 2 | 100 | 71 | |
BC152422.1 | - | 1 - 2 | 172 | 71 | |
D84223.1 | - | 1 - 2 | 77 | 71 | |
DB086913.1 | - | 1 - 2 | 244 | 71 |