Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
Chromosome Location
Chr.5: 150708747 - 150759095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135643.1 NP_001129115.1 12 - 13 1201 144
NM_001135644.1 NP_001129116.1 11 - 12 1348 144
NM_016221.3 NP_057305.1 11 - 12 1180 144
XM_005268457.1 XP_005268514.1 10 - 11 1074 144
XM_011537644.1 XP_011535946.1 12 - 13 1084 144
XM_011537645.1 XP_011535947.1 12 - 13 996 144
XM_011537646.1 XP_011535948.1 12 - 13 1217 144
XM_011537647.1 XP_011535949.1 12 - 13 1040 144
GenBank mRNA AF195120.1 - 11 - 12 1095 144
AK000299.1 - 11 - 12 1099 144
AK125973.1 - 11 - 12 1348 144
AK299517.1 - 11 - 12 144
AK307757.1 - 12 - 13 1140 144
AL162085.1 - 1 - 2 130 144
BC026323.1 - 11 - 12 1105 144
KU178637.1 - 11 - 12 1078 144
KU178639.1 - 7 - 8 834 144

View Full Product Details