Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SLC36A1
- Entrez Gene ID
-
206358
- Gene Name
- solute carrier family 36 member 1
- Gene Aliases
-
Dct1, LYAAT1, PAT1, TRAMD3
- Chromosome Location
-
Chr.5: 151411344 - 151556318 on Build GRCh38
View Full Product Details