The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_078483.3 | NP_510968.2 | 10 - 11 | 1376 | 101 |
XM_005268386.1 | XP_005268443.1 | 10 - 11 | 1346 | 101 | |
XM_006714759.3 | XP_006714822.1 | 10 - 11 | 1425 | 101 | |
XM_011537580.2 | XP_011535882.1 | 10 - 11 | 2066 | 101 | |
XM_011537581.1 | XP_011535883.1 | 11 - 12 | 1494 | 101 | |
XM_011537583.2 | XP_011535885.1 | 11 - 12 | 5931 | 101 | |
XM_011537584.2 | XP_011535886.1 | 10 - 11 | 1585 | 101 | |
XM_011537585.1 | XP_011535887.1 | 11 - 12 | 1441 | 101 | |
XM_011537586.2 | XP_011535888.1 | 10 - 11 | 2189 | 101 | |
XM_011537587.2 | XP_011535889.1 | 10 - 11 | 1445 | 101 | |
XM_011537588.2 | XP_011535890.1 | 10 - 11 | 1289 | 101 | |
XM_011537589.2 | XP_011535891.1 | 10 - 11 | 2197 | 101 | |
XM_011537590.1 | XP_011535892.1 | 10 - 11 | 1204 | 101 | |
XM_011537591.1 | XP_011535893.1 | 10 - 11 | 1179 | 101 | |
XM_011537592.2 | XP_011535894.1 | 10 - 11 | 1183 | 101 | |
XM_011537593.2 | XP_011535895.1 | 11 - 12 | 1439 | 101 | |
XM_011537594.1 | XP_011535896.1 | 12 - 13 | 1553 | 101 | |
XM_017009216.1 | XP_016864705.1 | 10 - 11 | 1272 | 101 | |
GenBank mRNA | AF516142.1 | - | 9 - 10 | 1168 | 101 |
AY162213.1 | - | 10 - 11 | 1281 | 101 | |
AY227112.1 | - | 4 - 5 | 664 | 101 | |
BC136437.1 | - | 10 - 11 | 1247 | 101 | |
BC136438.1 | - | 10 - 11 | 1247 | 101 | |
BX537963.1 | - | 10 - 11 | 1373 | 101 | |
GU296670.1 | - | 9 - 10 | 1153 | 101 |