The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008738.2 | NP_001008738.2 | 11 - 12 | 1405 | 110 |
NM_133372.2 | NP_588613.2 | 12 - 13 | 1489 | 110 | |
GenBank mRNA | AB075841.1 | - | 6 - 7 | 678 | 110 |
AK292473.1 | - | 12 - 13 | 1443 | 110 | |
BC001956.1 | - | 12 - 13 | 1442 | 110 | |
CR749426.1 | - | 6 - 7 | 728 | 110 | |
CR936723.1 | - | 11 - 12 | 1242 | 110 | |
DQ145719.1 | - | 12 - 13 | 1394 | 110 |