Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 131641714 - 131797063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133372.2 NP_588613.2 6 - 7 764 156
GenBank mRNA AK292473.1 - 6 - 7 718 156
BC001956.1 - 6 - 7 717 156
CR936723.1 - 5 - 6 517 156
DQ145719.1 - 6 - 7 669 156

View Full Product Details