Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 131641714 - 131797063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008738.2 NP_001008738.2 7 - 8 837 63
NM_133372.2 NP_588613.2 8 - 9 921 63
GenBank mRNA AB075841.1 - 2 - 3 110 63
AK292473.1 - 8 - 9 875 63
BC001956.1 - 8 - 9 874 63
CR749426.1 - 2 - 3 160 63
CR936723.1 - 7 - 8 674 63
DQ145719.1 - 8 - 9 826 63

View Full Product Details