Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 131641714 - 131797063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008738.2 NP_001008738.2 13 - 14 3003 63
NM_133372.2 NP_588613.2 14 - 15 3087 63
GenBank mRNA AB075841.1 - 8 - 9 2276 63
AK126675.1 - 1 - 2 447 63
AK292473.1 - 14 - 15 3041 63
AL832008.1 - 1 - 2 1099 63
AL834127.2 - 1 - 2 1165 63
CR749426.1 - 9 - 10 2303 63
CR936723.1 - 14 - 15 2817 63
DQ145719.1 - 14 - 15 2992 63

View Full Product Details