The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004395.3 | NP_004386.2 | 10 - 11 | 1134 | 64 |
NM_080881.2 | NP_543157.1 | 11 - 12 | 1250 | 64 | |
XM_017009140.1 | XP_016864629.1 | 10 - 11 | 1243 | 64 | |
GenBank mRNA | AK094125.1 | - | 9 - 10 | 1180 | 64 |
AK225523.1 | - | 10 - 11 | 1075 | 64 | |
AK295295.1 | - | 3 - 4 | 349 | 64 | |
AK314645.1 | - | 10 - 11 | 1075 | 64 | |
AL110225.1 | - | 11 - 12 | 1579 | 64 | |
BC000283.1 | - | 10 - 11 | 1058 | 64 | |
BC007281.1 | - | 10 - 11 | 1058 | 64 | |
BC007567.1 | - | 10 - 11 | 1058 | 64 | |
D17530.1 | - | 10 - 11 | 1059 | 64 | |
DR158741.1 | - | 1 - 2 | 218 | 64 | |
U00802.1 | - | 10 - 11 | 1034 | 64 |