Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBN1
Entrez Gene ID
1627
Gene Name
drebrin 1
Gene Aliases
D0S117E
Chromosome Location
Chr.5: 177456612 - 177480368 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004395.3 NP_004386.2 2 - 3 308 60
NM_080881.2 NP_543157.1 3 - 4 424 60
XM_005265827.3 XP_005265884.1 2 - 3 255 60
XM_006714826.2 XP_006714889.1 2 - 3 145 60
XM_011534446.2 XP_011532748.1 3 - 4 427 60
XM_011534447.2 XP_011532749.1 2 - 3 924 60
XM_017009139.1 XP_016864628.1 2 - 3 426 60
XM_017009140.1 XP_016864629.1 2 - 3 417 60
GenBank mRNA AK094125.1 - 2 - 3 239 60
AK225523.1 - 2 - 3 249 60
AK314645.1 - 2 - 3 249 60
AL110225.1 - 3 - 4 753 60
BC000283.1 - 2 - 3 232 60
BC007281.1 - 2 - 3 232 60
BC007567.1 - 2 - 3 232 60
BF205663.1 - - 280 60
D17530.1 - 2 - 3 233 60
DA245346.1 - 2 - 3 308 60
U00802.1 - 2 - 3 208 60

View Full Product Details