Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HK3
Entrez Gene ID
3101
Gene Name
hexokinase 3
Gene Aliases
HKIII, HXK3
Chromosome Location
Chr.5: 176880869 - 176899373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002115.2 NP_002106.2 10 - 11 1396 69
XM_011534540.2 XP_011532842.1 10 - 11 1437 69
XM_017009411.1 XP_016864900.1 9 - 10 1612 69
GenBank mRNA AB208951.1 - 8 - 9 2254 69
BC028129.1 - 10 - 11 1396 69
BX644742.1 - 1 - 2 335 69
U51333.1 - 10 - 11 1378 69

View Full Product Details