Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE2
Entrez Gene ID
84501
Gene Name
spire type actin nucleation factor 2
Gene Aliases
Spir-2
Chromosome Location
Chr.16: 89817477 - 89871319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032451.1 NP_115827.1 9 - 10 1512 96
XM_011523379.1 XP_011521681.1 9 - 10 2278 96
XM_011523380.2 XP_011521682.1 10 - 11 1534 96
XM_011523381.1 XP_011521683.1 10 - 11 2358 96
XM_011523384.1 XP_011521686.1 9 - 10 1536 96
XM_017023779.1 XP_016879268.1 9 - 10 1524 96
XM_017023780.1 XP_016879269.1 9 - 10 1689 96
XM_017023781.1 XP_016879270.1 8 - 9 1161 96
GenBank mRNA AB058735.1 - 7 - 8 1610 96
AJ422077.1 - 9 - 10 1502 96
AK094918.1 - 2 - 3 329 96
AL834408.1 - 9 - 10 1306 96
BC063706.1 - 1 - 2 125 96
BC111030.1 - 9 - 10 1512 96
BC139732.1 - 7 - 8 1130 96

View Full Product Details