The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024870.3 | NP_079146.2 | 24 - 25 | 3215 | 102 |
XM_011517612.2 | XP_011515914.1 | 24 - 25 | 3409 | 102 | |
GenBank mRNA | AJ437636.2 | - | 24 - 25 | 3212 | 102 |
AK024079.1 | - | 9 - 10 | 1291 | 102 | |
AK294299.1 | - | 22 - 23 | 2646 | 102 | |
AY508996.1 | - | 24 - 25 | 2962 | 102 |