The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024870.3 | NP_079146.2 | 26 - 27 | 3611 | 67 |
XM_011517612.2 | XP_011515914.1 | 26 - 27 | 3805 | 67 | |
GenBank mRNA | AJ437636.2 | - | 26 - 27 | 3608 | 67 |
AK024079.1 | - | 11 - 12 | 1687 | 67 | |
AY508996.1 | - | 26 - 27 | 3358 | 67 |