The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017813.4 | NP_060283.3 | 2 - 3 | 837 | 72 |
GenBank mRNA | AK000428.1 | - | 2 - 3 | 834 | 72 |
AY032885.1 | - | 2 - 3 | 551 | 72 | |
BC017797.1 | - | 2 - 3 | 786 | 72 | |
BC067814.1 | - | 2 - 3 | 810 | 72 |