The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303.3 | NP_001294.2 | 2 - 3 | 374 | 118 |
GenBank mRNA | AK295925.1 | - | 2 - 3 | 172 | 118 |
AK312718.1 | - | 2 - 3 | 269 | 118 | |
BC000060.2 | - | 2 - 3 | 247 | 118 | |
BC006394.2 | - | 2 - 3 | 247 | 118 | |
BT006985.1 | - | 2 - 3 | 170 | 118 | |
U09466.1 | - | 2 - 3 | 210 | 118 |