The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018064.2 | NP_001018074.1 | 12 - 13 | 1950 | 152 |
NM_006180.4 | NP_006171.2 | 16 - 17 | 2657 | 152 | |
XM_005252001.2 | XP_005252058.1 | 15 - 16 | 2538 | 152 | |
XM_005252003.2 | XP_005252060.1 | 15 - 16 | 2354 | 152 | |
XM_005252004.2 | XP_005252061.1 | 14 - 15 | 2254 | 152 | |
XM_011518718.2 | XP_011517020.1 | 14 - 15 | 2490 | 152 | |
XM_017014751.1 | XP_016870240.1 | 16 - 17 | 3080 | 152 | |
XM_017014752.1 | XP_016870241.1 | 15 - 16 | 2545 | 152 | |
XM_017014753.1 | XP_016870242.1 | 15 - 16 | 3032 | 152 | |
XM_017014754.1 | XP_016870243.1 | 13 - 14 | 2206 | 152 | |
GenBank mRNA | AB209118.1 | - | 15 - 16 | 2511 | 152 |
AF400441.1 | - | 12 - 13 | 1683 | 152 | |
AF410899.1 | - | 14 - 15 | 2108 | 152 | |
S76473.1 | - | 12 - 13 | 1691 | 152 | |
U12140.1 | - | 12 - 13 | 1929 | 152 |