The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018064.2 | NP_001018074.1 | 15 - 16 | 2497 | 97 |
NM_006180.4 | NP_006171.2 | 19 - 20 | 3204 | 97 | |
XM_005252001.2 | XP_005252058.1 | 18 - 19 | 3085 | 97 | |
XM_005252003.2 | XP_005252060.1 | 18 - 19 | 2901 | 97 | |
XM_005252004.2 | XP_005252061.1 | 17 - 18 | 2801 | 97 | |
XM_011518718.2 | XP_011517020.1 | 17 - 18 | 3037 | 97 | |
XM_017014751.1 | XP_016870240.1 | 19 - 20 | 3627 | 97 | |
XM_017014752.1 | XP_016870241.1 | 18 - 19 | 3092 | 97 | |
XM_017014753.1 | XP_016870242.1 | 18 - 19 | 3579 | 97 | |
XM_017014754.1 | XP_016870243.1 | 16 - 17 | 2753 | 97 | |
GenBank mRNA | AF400441.1 | - | 15 - 16 | 2230 | 97 |
AF410899.1 | - | 17 - 18 | 2655 | 97 | |
S76473.1 | - | 15 - 16 | 2238 | 97 | |
U12140.1 | - | 15 - 16 | 2476 | 97 |