The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278241.1 | NP_001265170.1 | 10 - 11 | 1379 | 89 |
NM_014389.2 | NP_055204.3 | 9 - 10 | 1297 | 89 | |
GenBank mRNA | AF547989.1 | - | 9 - 10 | 1071 | 89 |
AK226056.1 | - | 9 - 10 | 1077 | 89 | |
AK291163.1 | - | 9 - 10 | 1097 | 89 | |
AK293838.1 | - | 10 - 11 | 1166 | 89 | |
AK299084.1 | - | 9 - 10 | 1285 | 89 | |
AY882602.1 | - | 9 - 10 | 864 | 89 | |
BC010457.2 | - | 8 - 9 | 825 | 89 | |
BC069058.1 | - | 9 - 10 | 1071 | 89 |